(+)-Menthyl acetate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L13987-03
L13987-MD
CAAAAL13987-03EA
133.67
CAD
CAAAAL13987-03
CAAAAL13987-MD
(+)-Menthyl acetate 99%
(+)-Menthyl acetate
Purity: 99%
CAS No.: 5157-89-1
Molecular Formula: C12H22O2
Formula Weight: 198.31
Flash Point: 77°(170°F)
Refractive Index: 1.4470
Boiling Point: 229-230°
Beilstein Reference: 1909419
Optical Rotation: +80.5° (c=8 in benzene)
MDL No.: MFCD00062977
Formula:
C₁₂H₂₂O₂ Boiling Pt: 229…230 °C Density: 0.925 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00062977 CAS Number: 5157-89-1 |
Specification Test Results
Optical rotation | +80.5° (c=8 in benzene) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...