Sodium-D(+)-glucuronate monohydrate 99%
Supplier: Thermo Scientific Chemicals
L13985-06
L13985-14
CAAAAL13985-06EA
46.29
CAD
CAAAAL13985-06
CAAAAL13985-14
Sodium-D(+)-glucuronate monohydrate 99%
Sodium D(+)-glucuronate monohydrate
Purity: 99%
CAS No.: 207300-70-7
Molecular Formula: C6H9NaO7.H2O
Formula Weight: 234.14 (216.14anhy)
Melting Point: 134-138°
Beilstein Reference: 4218577
Formula:
C₆H₁₀O₇·1H₂O Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151051 CAS Number: 207300-70-7 EINECS: 239-065-7 |
Specification Test Results
Optical rotation | +21° (c=2 in water, 2h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...