Benzyl (R)-(-)-glycidyl ether 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-2-((Benzyloxy)methyl)oxirane

L13980-03 L13980-06
CAAAAL13980-03EA 124.03 CAD
CAAAAL13980-03 CAAAAL13980-06
Benzyl (R)-(-)-glycidyl ether 98+%
Benzyl (R)-(-)-glycidyl ether

Purity: 98+% CAS No.: 14618-80-5 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 130°/0.1mm Beilstein Reference: 3588399 Optical Rotation: -5.1° (c=5 in toluene) MDL No.: MFCD00068409

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Boiling Pt: 130 °C/0.1 mm
Density: 1.072
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00068409
CAS Number: 14618-80-5

Order Now

Specification Test Results

Optical rotation -5.1° (c=5 in toluene)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR