N-Fluorobenzenesulfonimide 97%

Supplier: Thermo Scientific Chemicals
Warning

L13955-03 L13955-06 L13955-14
CAAAAL13955-03EA 48.51 CAD
CAAAAL13955-03 CAAAAL13955-06 CAAAAL13955-14
N-Fluorobenzenesulfonimide 97%
N-Fluorobenzenesulphonimide

Fieser: 18,161 20,165 21,189


Purity: 97% CAS No.: 133745-75-2 Molecular Formula: C12H10FNO4S2 Formula Weight: 315.34 Melting Point: 114-118° Beilstein Reference: 5348902

Formula: C₁₂H₁₀FNO₄S₂
MW: 315.34 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00144885
CAS Number: 133745-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR