Dowex® 50WX2 (H), Dowex® 200-400 mesh

Supplier: Thermo Scientific Chemicals
Danger

L13945-0B L13945-18 L13945-30
CAAAAL13945-0BEA 1327.2 CAD
CAAAAL13945-0B CAAAAL13945-18 CAAAAL13945-30
Dowex® 50WX2 (H), Dowex® 200-400 mesh
Dowex® 50WX2 (H)

CAS No.: 12612-37-2 MDL No.: MFCD00132722

Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 12612-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR