Dowex® 50WX2 (H), Dowex® 100-200 mesh

Supplier: Thermo Scientific Chemicals
Danger

L13944-0B L13944-18 L13944-30
CAAAAL13944-0BEA 843.72 CAD
CAAAAL13944-0B CAAAAL13944-18 CAAAAL13944-30
Dowex® 50WX2 (H), Dowex® 100-200 mesh
Dowex® 50WX2 (H)

Fieser: 21,236


CAS No.: 12612-37-2 MDL No.: MFCD00132722

Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 12612-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR