Dowex® 50WX2 (H), Dowex® 50-100 mesh

Supplier: Thermo Scientific Chemicals
Danger

L13943-0B L13943-18 L13943-30
CAAAAL13943-0BEA 955.9 CAD
CAAAAL13943-0B CAAAAL13943-18 CAAAAL13943-30
Dowex® 50WX2 (H), Dowex® 50-100 mesh
Dowex® 50WX2 (H)

Fieser: 1,511 4,266 5,355 6,302 9,256 18,194


CAS No.: 12612-37-2 MDL No.: MFCD00132722

Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 12612-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR