Dowex® 50WX8 (H), Dowex® 200-400 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dowex 50 wx8
L13922-0B
L13922-18
L13922-30
CAAAAL13922-0BEA
1077.56
CAD
CAAAAL13922-0B
CAAAAL13922-18
CAAAAL13922-30
Dowex® 50WX8 (H), Dowex® 200-400 mesh
Dowex® 50WX8 (H)
CAS No.: 11119-67-8
MDL No.: MFCD00132726
Storage Temperature:
Ambient |
MDL Number:
MFCD00132726 CAS Number: 11119-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...