Dowex® 50WX8 (H), Dowex® 100-200 mesh

Supplier: Thermo Scientific Chemicals
Danger

L13921-0B L13921-18 L13921-30
CAAAAL13921-0BEA 1339.84 CAD
CAAAAL13921-0B CAAAAL13921-18 CAAAAL13921-30
Dowex® 50WX8 (H), Dowex® 100-200 mesh
Dowex® 50WX8 (H)

CAS No.: 11119-67-8 MDL No.: MFCD00132726

Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 11119-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR