Dowex® 50WX4 (H), Dowex® 100-200 mesh

Supplier: Thermo Scientific Chemicals
Danger

L13917-0B L13917-18 L13917-30
CAAAAL13917-0BEA 1094.94 CAD
CAAAAL13917-0B CAAAAL13917-18 CAAAAL13917-30
Dowex® 50WX4 (H), Dowex® 100-200 mesh
Dowex® 50WX4 (H)

CAS No.: 11113-61-4 MDL No.: MFCD00132725

Storage Temperature: Ambient
MDL Number: MFCD00132725
CAS Number: 11113-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR