2-Formylimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Imidazole-2-carbaldehyde, Imidazole-2-carboxaldehyde

L13883-06 L13883-14
CAAAAL13883-06EA 176.96 CAD
CAAAAL13883-06 CAAAAL13883-14
2-Formylimidazole 97%
2-Formylimidazole

Purity: 97% CAS No.: 10111-08-7 Molecular Formula: C4H4N2O Formula Weight: 96.09 Storage Temperature: 2°C to 8°C Melting Point: ca 205° dec. Beilstein Reference: 4371302 MDL No.: MFCD00003544

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003544
CAS Number: 10111-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR