2-Amino-4-(p-tolyl)thiazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(p-Tolyl)-2-thiazolamine, 4-(4-Methylphenyl)-2-thiazolamine, 4-(p-Tolyl)thiazol-2-amine, 2-Amino-4-(4-methylphenyl)thiazole
L13863-03
L13863-06
CAAAAL13863-03EA
42.98
CAD
CAAAAL13863-03
CAAAAL13863-06
2-Amino-4-(p-tolyl)thiazole 98%
2-Amino-4-(p-tolyl)thiazole
Purity: 98%
CAS No.: 2103-91-5
Molecular Formula: C10H10N2S
Formula Weight: 190.27
Melting Point: 134-138°
Beilstein Reference: 143563
MDL No.: MFCD00170264
Formula:
C₁₀H₁₀N₂S MW: 190.26 g/mol Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00170264 CAS Number: 2103-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...