N-(Ethoxycarbonyl)nortropinone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one, N-Carbethoxy-4-tropinone

L13849-06 L13849-14
CAAAAL13849-03EA 56.96 CAD
CAAAAL13849-03 CAAAAL13849-06 CAAAAL13849-14
N-(Ethoxycarbonyl)nortropinone 99%
N-(Ethoxycarbonyl)nortropinone

Purity: 99% CAS No.: 32499-64-2 Molecular Formula: C10H15NO3 Formula Weight: 197.23 Refractive Index: 1.4895 Boiling Point: 152-153°/11mm Beilstein Reference: 1530944 MDL No.: MFCD00078171

Formula: C₁₀H₁₅NO₃
MW: 197.23 g/mol
Boiling Pt: 152…153 °C/11 mm
Storage Temperature: Ambient
MDL Number: MFCD00078171
CAS Number: 32499-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR