(±)-tert-Butyl-2-bromobutyrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13799-06
L13799-14
CAAAAL13799-06EA
51.82
CAD
CAAAAL13799-06
CAAAAL13799-14
(±)-tert-Butyl-2-bromobutyrate 98%
(±)-tert-Butyl 2-bromobutyrate
Purity: 98%
CAS No.: 24457-21-4
Molecular Formula: C8H15BrO2
Formula Weight: 223.12
Refractive Index: 1.4410
Boiling Point: 72-76°/12mm
UN No.: 1760
MDL No.: MFCD00209634
Formula:
C₈H₁₅BrO₂ Boiling Pt: 72…76 °C/12 mm Density: 1.200 Storage Temperature: Ambient |
MDL Number:
MFCD00209634 CAS Number: 24457-21-4 UN: 1760 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...