4'-(4-Bromophenyl)acetophenone 97%
Supplier: Thermo Scientific Chemicals
L13798-14
L13798-22
CAAAAL13798-14EA
114.55
CAD
CAAAAL13798-14
CAAAAL13798-22
4'-(4-Bromophenyl)acetophenone 97%
4'-(4-Bromophenyl)acetophenone
Purity: 97%
CAS No.: 5731-01-1
Molecular Formula: C14H11BrO
Formula Weight: 275.15
Melting Point: 125-127°
Beilstein Reference: 2048590
Formula:
C₁₄H₁₁BrO Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143242 CAS Number: 5731-01-1 EINECS: 227-236-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...