Diisopropyl bromomethylphosphonate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13763-06
L13763-14
CAAAAL13763-06EA
148.05
CAD
CAAAAL13763-06
CAAAAL13763-14
Diisopropyl bromomethylphosphonate 97%
Diisopropyl bromomethylphosphonate
Purity: 97%
CAS No.: 98432-80-5
Molecular Formula: C7H16BrO3P
Formula Weight: 259.09
Refractive Index: 1.4495
Boiling Point: 80°/1mm
Beilstein Reference: 4740388
UN No.: 3265
MDL No.: MFCD00236036
Formula:
C₇H₁₆BrO₃P Boiling Pt: 80 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00236036 CAS Number: 98432-80-5 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...