3-Hydroxy-o-toluic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13743-03
L13743-06
CAAAAL13743-03EA
51.67
CAD
CAAAAL13743-03
CAAAAL13743-06
3-Hydroxy-o-toluic acid 97%
Purity: 97%
CAS No.: 603-80-5
Molecular Formula: C8H8O3
Formula Weight: 152.15
Odor: Odorless
Melting Point: 142-146°
Beilstein Reference: 2613791
MDL No.: MFCD00671541
Formula:
C₈H₈O₃ Melting Pt: 142…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671541 CAS Number: 603-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...