1-Bromo-4-(ethylthio)benzene 97%
Supplier: Thermo Scientific Chemicals
L13718-06
L13718-14
L13718-22
CAAAAL13718-06EA
51.98
CAD
CAAAAL13718-06
CAAAAL13718-14
CAAAAL13718-22
1-Bromo-4-(ethylthio)benzene 97%
1-Bromo-4-(ethylthio)benzene
Purity: 97%
CAS No.: 30506-30-0
Molecular Formula: C8H9BrS
Formula Weight: 217.13
Refractive Index: 1.6069
Boiling Point: 109-111°/2mm
MDL No.: MFCD01318109
Formula:
C₈H₉BrS MW: 217.13 g/mol Boiling Pt: 109…111 °C/2 mm Storage Temperature: Ambient |
MDL Number:
MFCD01318109 CAS Number: 30506-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...