2-Amino-5-n-propylsulfonylbenzimidazole ≥98%

Supplier: Thermo Scientific Chemicals

L13711-03 L13711-MC
CAAAAL13711-03EA 489.8 CAD
CAAAAL13711-03 CAAAAL13711-MC
2-Amino-5-n-propylsulfonylbenzimidazole ≥98%
2-Amino-5-n-propylsulfonylbenzimidazole

Purity: 98+% CAS No.: 80983-34-2 Molecular Formula: C10H13N3O2S Formula Weight: 239.30 Melting Point: 220-222° MDL No.: MFCD01075656

Formula: C₁₀H₁₃N₃O₂S
Melting Pt: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD01075656
CAS Number: 80983-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR