3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13709-03
L13709-06
CAAAAL13709-03EA
123.08
CAD
CAAAAL13709-03
CAAAAL13709-06
CAAAAL13709-MD
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane 95%
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
Purity: 95%
CAS No.: 732306-23-9
Molecular Formula: C10H9ClF6Si
Formula Weight: 306.71
Refractive Index: 1.4240
Boiling Point: 95-96°/30mm
UN No.: 3265
MDL No.: MFCD01862186
Formula:
C₁₀H₉ClF₆Si Boiling Pt: 95…96 °C/30 mm Storage Temperature: Ambient |
MDL Number:
MFCD01862186 CAS Number: 732306-23-9 UN: 2987 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...