5-Acetylthiophene-2-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Acetylthiophene-2-carboxylic Acid, 5-Acetyl-2-thenoic acid

L13588-06 L13588-14
CAAAAL13588-06EA 122.29 CAD
CAAAAL13588-06 CAAAAL13588-14
5-Acetylthiophene-2-carboxylic acid 98+%
5-Acetylthiophene-2-carboxylic acid

Purity: 98+% CAS No.: 4066-41-5 Molecular Formula: C7H6O3S Formula Weight: 170.19 Melting Point: 208-212° Beilstein Reference: 128897

Formula: C₇H₆O₃S
MW: 170.19 g/mol
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00055512
CAS Number: 4066-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR