5-Bromo-3-pyridineacetic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13579-03
L13579-06
CAAAAL13579-03EA
173.8
CAD
CAAAAL13579-03
CAAAAL13579-06
5-Bromo-3-pyridineacetic acid ≥98%
5-Bromo-3-pyridineacetic acid
Purity: 98+%
CAS No.: 39891-12-8
Molecular Formula: C7H6BrNO2
Formula Weight: 216.03
Melting Point: 182-185°
Beilstein Reference: 388264
MDL No.: MFCD00829308
Formula:
C₇H₆BrNO₂ Melting Pt: 182…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00829308 CAS Number: 39891-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...