Methyl-2-methoxypropionate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13527-06
L13527-14
CAAAAL13527-06EA
70.78
CAD
CAAAAL13527-06
CAAAAL13527-14
Methyl-2-methoxypropionate 98%
Methyl-2-methoxypropionate
Purity: 98%
CAS No.: 17639-76-8
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 36°(96°F)
Refractive Index: 1.3970
Boiling Point: 43-44°/15mm
Beilstein Reference: 1720819
UN No.: 3272
MDL No.: MFCD00151664
Formula:
C₅H₁₀O₃ Boiling Pt: 43…44 °C/15 mm Flash Pt: 36 °C(96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151664 CAS Number: 17639-76-8 EINECS: 241-622-4 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...