Methacroylcholine chloride ≥72% in aqueous solution stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Methacryloyloxy)-N,N,N-trimethylethanaminium chloride, (Methacryloyloxyethyl)trimethyl ammonium chloride, 2-(Methacryloyloxy)ethyltrimethylammonium chloride, Trimethyl-2-methacroyloxyethylammonium Chloride

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 150-200 ppm

L13523-22 L13523-36
CAAAAL13523-22EA 47.87 CAD
CAAAAL13523-22 CAAAAL13523-36
Methacroylcholine chloride ≥72% in aqueous solution stabilized
Methacroylcholine chloride

CAS No.: 5039-78-1 Molecular Formula: C9H18ClNO2 Formula Weight: 207.70 Refractive Index: 1.4660 Melting Point: -25° Boiling Point: 100° Beilstein Reference: 8168395 MDL No.: MFCD00060097

Formula: C₉H₁₈ClNO₂
MW: 207.70 g/mol
Boiling Pt: >100 °C
Melting Pt: –25 °C
Density: 1.105
Storage Temperature: Ambient
MDL Number: MFCD00060097
CAS Number: 5039-78-1
EINECS: 225-733-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR