2-(Dimethylamino)ethyl acrylate 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 0.1%
L13506-AC
L13506-AE
L13506-AP
CAAAAL13506-ACEA
43.45
CAD
CAAAAL13506-AC
CAAAAL13506-AE
CAAAAL13506-AP
2-(Dimethylamino)ethyl acrylate 98% stabilized
2-(Dimethylamino)ethyl acrylate
Stab. with ca 0.1% 4-methoxyphenol
Purity: 98%
CAS No.: 2439-35-2
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Storage Temperature: 2°C to 8°C
Flash Point: 62°(144°F)
Refractive Index: 1.4380
Melting Point: -60°
Boiling Point: 64°/12mm
Beilstein Reference: 1099119
UN No.: 3302
MDL No.: MFCD00038233
Formula:
C₇H₁₃NO₂ Boiling Pt: 64 °C/12 mm Melting Pt: –60 °C Density: 0.943 Flash Pt: 62 °C(144 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00038233 CAS Number: 2439-35-2 EINECS: 219-460-0 UN: 3302 ADR: "6.1",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...