Boc-sarcosine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-Sar-OH

L13482-06 L13482-14
CAAAAL13482-06EA 73.47 CAD
CAAAAL13482-06 CAAAAL13482-14
Boc-sarcosine 99%
Boc-sarcosine

Purity: 99% CAS No.: 13734-36-6 Molecular Formula: C8H15NO4 Formula Weight: 189.20 Storage Temperature: 2°C to 8°C Melting Point: 88-93° Beilstein Reference: 2046827 MDL No.: MFCD00037795

Formula: C₈H₁₅NO₄
MW: 189.211 g/mol
Melting Pt: 88…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037795
CAS Number: 13734-36-6
EINECS: 237-306-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR