TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TBTU

L13470-03 L13470-06 L13470-14
CAAAAL13470-03EA 41.55 CAD
CAAAAL13470-03 CAAAAL13470-06 CAAAAL13470-14
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 99%
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)

Purity: 99% CAS No.: 125700-67-6 Molecular Formula: C11H16BF4N5O Formula Weight: 321.08 Melting Point: 195-198° dec. Beilstein Reference: 7066325

Formula: C₁₁H₁₆BF₄N₅O
MW: 321.1 g/mol
Melting Pt: 195…198 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00077413
CAS Number: 125700-67-6
EINECS: 423-040-4
UN: 1325
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR