4,4'-Diphenoxybenzophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(4-phenoxyphenyl)methanone
L13419-03
L13419-06
L13419-14
CAAAAL13419-03EA
47.87
CAD
CAAAAL13419-03
CAAAAL13419-06
CAAAAL13419-14
4,4'-Diphenoxybenzophenone 98%
4,4'-Diphenoxybenzophenone
Purity: 98%
CAS No.: 14984-21-5
Molecular Formula: C25H18O3
Formula Weight: 366.42
Melting Point: 147-151°
Beilstein Reference: 2165449
Formula:
C₂₅H₁₈O₃ Melting Pt: 147…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077969 CAS Number: 14984-21-5 EINECS: 403-390-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...