1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13407-03
L13407-06
L13407-14
CAAAAL13407-03EA
52.3
CAD
CAAAAL13407-03
CAAAAL13407-06
CAAAAL13407-14
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) 96%
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)
Purity: 96%
CAS No.: 2549-93-1
Molecular Formula: C8H18N2
Formula Weight: 142.25
Flash Point: 106°(222°F)
Refractive Index: 1.4900
Boiling Point: 239-244°
Beilstein Reference: 2071383
UN No.: 2922
MDL No.: MFCD00039463
Formula:
C₈H₁₈N₂ Boiling Pt: 239…244 °C Density: 0.950 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039463 CAS Number: 2549-93-1 EINECS: 219-840-6 UN: 3267 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...