1-Pyrrolidinecarbonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13396-03
L13396-06
CAAAAL13396-03EA
59.72
CAD
CAAAAL13396-03
CAAAAL13396-06
1-Pyrrolidinecarbonyl chloride 97%
1-Pyrrolidinecarbonyl chloride
Purity: 97%
CAS No.: 1192-63-8
Molecular Formula: C5H8ClNO
Formula Weight: 133.58
Storage Temperature: 2°C to 8°C
Flash Point: 97°(206°F)
Refractive Index: 1.4940
Boiling Point: 75°/1mm
Beilstein Reference: 110281
UN No.: 3265
MDL No.: MFCD00051321
Formula:
C₅H₈ClNO Boiling Pt: 75 °C/1 mm Density: 1.209 Flash Pt: 97 °C(206 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00051321 CAS Number: 1192-63-8 EINECS: 214-758-7 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...