N-tert-Butyl-2-benzothiazolesulfenamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13356-18
L13356-30
CAAAAL13356-18EA
48.98
CAD
CAAAAL13356-18
CAAAAL13356-30
N-tert-Butyl-2-benzothiazolesulfenamide 97%
N-tert-Butyl-2-benzothiazolesulfenamide
Purity: 97%
CAS No.: 95-31-8
Molecular Formula: C11H14N2S2
Formula Weight: 238.38
Flash Point: 165°(329°F)
Melting Point: 105-110°
Beilstein Reference: 158370
MDL No.: MFCD00022873
Formula:
C₁₁H₁₄N₂S₂ Melting Pt: 105…110 °C Density: 1.290 Flash Pt: 165 °C(329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022873 CAS Number: 95-31-8 EINECS: 202-409-1 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...