Cyclopropanemethylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13353-03
L13353-06
CAAAAL13353-03EA
69.52
CAD
CAAAAL13353-03
CAAAAL13353-06
Cyclopropanemethylamine hydrochloride 98%
Cyclopropanemethylamine hydrochloride
Purity: 98%
CAS No.: 7252-53-1
Molecular Formula: C4H9N.HCl
Formula Weight: 107.58
Melting Point: 202-205°
Beilstein Reference: 3906936
MDL No.: MFCD00012544
Formula:
C₄H₉N·HCl Melting Pt: 202…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012544 CAS Number: 7252-53-1 EINECS: 230-668-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...