2-(Hydroxymethyl)-2-nitro-1,3-propanediol 95%

Supplier: Thermo Scientific Chemicals
Warning

L13349-22 L13349-36
CAAAAL13349-22EA 63.36 CAD
CAAAAL13349-22 CAAAAL13349-36
2-(Hydroxymethyl)-2-nitro-1,3-propanediol 95%
2-(Hydroxymethyl)-2-nitro-1,3-propanediol

Purity: 95% CAS No.: 126-11-4 Molecular Formula: C4H9NO5 Formula Weight: 151.12 Melting Point: ca 160° dec. Beilstein Reference: 742427 MDL No.: MFCD00007395

Formula: C₄H₉NO₅
Melting Pt: ca 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007395
CAS Number: 126-11-4
EINECS: 204-769-5
Merck Index: 14,9753

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR