4,4'-Biphenyldiboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
[1,1'-Biphenyl]-4,4'-diyldiboronic acid
L13328-03
L13328-06
CAAAAL13328-03EA
371.3
CAD
CAAAAL13328-03
CAAAAL13328-06
4,4'-Biphenyldiboronic acid 95%
4,4'-Biphenyldiboronic acid
Purity: 95%
CAS No.: 4151-80-8
Molecular Formula: C12H12B2O4
Formula Weight: 241.85
Melting Point: ca 300° dec.
Beilstein Reference: 2941876
Formula:
C₁₂H₁₂B₂O₄ MW: 241.85 g/mol Melting Pt: ca 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00151795 CAS Number: 4151-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...