(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione ≥98%
Supplier: Thermo Scientific Chemicals
L13326-03
L13326-06
CAAAAL13326-03EA
297.04
CAD
CAAAAL13326-03
CAAAAL13326-06
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione ≥98%
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione
Purity: 98+%
CAS No.: 20007-72-1
Molecular Formula: C11H14O2
Formula Weight: 178.23
Storage Temperature: 2°C to 8°C
Melting Point: 47-50°
Beilstein Reference: 3201286
MDL No.: MFCD00001687
Formula:
C₁₁H₁₄O₂ Melting Pt: 47…50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001687 CAS Number: 20007-72-1 EINECS: 243-463-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...