Allyl vinyl ether 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with potassium hydroxide 0.1%

L13320-09 L13320-18
CAAAAL13320-09EA 153.1 CAD
CAAAAL13320-09 CAAAAL13320-18
Allyl vinyl ether 95% stabilized
Allyl vinyl ether

Stab. with 0.1% potassium hydroxide


Purity: 95% CAS No.: 3917-15-5 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: -5°(23°F) Refractive Index: 1.4120 Odor: Characteristic Boiling Point: 65-68° Beilstein Reference: 1697665 UN No.: 3271 MDL No.: MFCD00039827

Formula: C₅H₈O
Boiling Pt: 65…68 °C
Density: 0.781
Flash Pt: <-5 °C(23 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039827
CAS Number: 3917-15-5
EINECS: 223-482-6
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR