N-Carbobenzoxyhydroxylamine ≥98%
Supplier: Thermo Scientific Chemicals
L13175-06
L13175-14
CAAAAL13175-06EA
135.25
CAD
CAAAAL13175-06
CAAAAL13175-14
N-Carbobenzoxyhydroxylamine ≥98%
N-Carbobenzoxyhydroxylamine
Purity: 98+%
CAS No.: 3426-71-9
Molecular Formula: C6H5CH2OCONHOH
Formula Weight: 167.17
Melting Point: 65-70°
Beilstein Reference: 1074034
MDL No.: MFCD00010642
Formula:
C₈H₉NO₃ Melting Pt: 65…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010642 CAS Number: 3426-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...