1-(Triisopropylsilyl)pyrrole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(Triisopropylsilyl)-1H-pyrrole
L13152-06
L13152-14
CAAAAL13152-03EA
93.57
CAD
CAAAAL13152-03
CAAAAL13152-06
CAAAAL13152-14
1-(Triisopropylsilyl)pyrrole 95%
1-(Triisopropylsilyl)pyrrole
Purity: 95%
CAS No.: 87630-35-1
Molecular Formula: C13H25NSi
Formula Weight: 223.44
Flash Point: 106°(222°F)
Refractive Index: 1.4920
Boiling Point: 78°/0.4mm
Beilstein Reference: 3540663
MDL No.: MFCD00054932
Formula:
C₁₃H₂₅NSi MW: 223.44 g/mol Boiling Pt: 78 °C/0.4 mm Density: 0.904 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00054932 CAS Number: 87630-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...