1-Phenyl-1,4-pentanedione 96%
Supplier: Thermo Scientific Chemicals
L13151-03
L13151-06
CAAAAL13151-03EA
69.52
CAD
CAAAAL13151-03
CAAAAL13151-06
1-Phenyl-1,4-pentanedione 96%
1-Phenyl-1,4-pentanedione
Purity: 96%
CAS No.: 583-05-1
Molecular Formula: C11H12O2
Formula Weight: 176.22
Refractive Index: 1.5330
Boiling Point: 158-162°/12mm
Beilstein Reference: 908230
MDL No.: MFCD00092889
Formula:
C₁₁H₁₂O₂ Boiling Pt: 158…162 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD00092889 CAS Number: 583-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...