L-Leucine ethyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals

L13147-06 L13147-14
CAAAAL13147-06EA 62.41 CAD
CAAAAL13147-06 CAAAAL13147-14
L-Leucine ethyl ester hydrochloride 97%
L-Leucine ethyl ester hydrochloride

Purity: 97% CAS No.: 2743-40-0 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Odor: Odorless Melting Point: 134-137° Beilstein Reference: 3994312 Optical Rotation: +18° (c=5 in ethanol) MDL No.: MFCD00034879

Formula: C₈H₁₇NO₂
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00034879
CAS Number: 2743-40-0
EINECS: 220-375-6

Order Now

Specification Test Results

Optical rotation +18° (c=5 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR