Ethyl mesitylglyoxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13106-06
L13106-14
CAAAAL13106-06EA
72.52
CAD
CAAAAL13106-06
CAAAAL13106-14
Ethyl mesitylglyoxylate 97%
Ethyl mesitylglyoxylate
Purity: 97%
CAS No.: 5524-57-2
Molecular Formula: C13H16O3
Formula Weight: 220.27
Refractive Index: 1.5120
Boiling Point: 164-165°/10mm
Beilstein Reference: 2970031
MDL No.: MFCD00151821
Formula:
C₁₃H₁₆O₃ Boiling Pt: 164…165 °C/10 mm Storage Temperature: Ambient |
MDL Number:
MFCD00151821 CAS Number: 5524-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...