2,6-Dibromo-4-nitrotoluene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13102-06
L13102-14
CAAAAL13102-06EA
90.38
CAD
CAAAAL13102-06
CAAAAL13102-14
2,6-Dibromo-4-nitrotoluene 95%
2,6-Dibromo-4-nitrotoluene
Purity: 95%
CAS No.: 110127-07-6
Molecular Formula: C7H5Br2NO2
Formula Weight: 294.94
Melting Point: 54-57°
Beilstein Reference: 3259701
Formula:
C₇H₅Br₂NO₂ Melting Pt: 54…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151807 CAS Number: 110127-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...