3,5-Dibromo-p-toluidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dibromo-4-methylaniline
L13101-03
L13101-06
CAAAAL13101-03EA
101.91
CAD
CAAAAL13101-03
CAAAAL13101-06
3,5-Dibromo-p-toluidine 99%
3,5-Dibromo-p-toluidine
Purity: 99%
CAS No.: 13194-73-5
Molecular Formula: C7H7Br2N
Formula Weight: 264.96
Melting Point: 90-92°
Beilstein Reference: 2639285
Formula:
C₇H₇Br₂N MW: 264.96 g/mol Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151806 CAS Number: 13194-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...