Benzoyl-DL-alanine ≥97%
Supplier: Thermo Scientific Chemicals
L13100-09
L13100-18
CAAAAL13100-09EA
63.67
CAD
CAAAAL13100-09
CAAAAL13100-18
Benzoyl-DL-alanine ≥97%
Benzoyl-DL-alanine
Purity: 97+%
CAS No.: 1205-02-3
Molecular Formula: C10H11NO3
Formula Weight: 193.20
Melting Point: 163-167°
Beilstein Reference: 3201778
MDL No.: MFCD00020393
Formula:
C₁₀H₁₁NO₃ Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020393 CAS Number: 1205-02-3 EINECS: 214-879-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...