Ammeline 90%, tech.

Supplier: Thermo Scientific Chemicals

Synonyms: Cyanurodiamide, 4,6-Diamino-1,3,5-triazin-2-ol, 2,4-Diamino-6-hydroxy-1,3,5-triazine, 3,4,5,6-Tetrahydro-4,6-diimino-1,3,5-triazin-2(1H)-one

L13064-03 L13064-06
CAAAAL13064-03EA 83.11 CAD
CAAAAL13064-03 CAAAAL13064-06
Ammeline 90%, tech.
Ammeline

Purity: 90% CAS No.: 645-92-1 Molecular Formula: C3H5N5O Formula Weight: 127.11 Beilstein Reference: 5570 MDL No.: MFCD00023186

Formula: C₃H₅N₅O
MW: 127.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023186
CAS Number: 645-92-1
EINECS: 211-455-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR