4-(1-Bromoethyl)benzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13025-03
L13025-06
CAAAAL13025-03EA
39.03
CAD
CAAAAL13025-03
CAAAAL13025-06
4-(1-Bromoethyl)benzoic acid 98%
4-(1-Bromoethyl)benzoic acid
Purity: 98%
CAS No.: 113023-73-7
Molecular Formula: C9H9BrO2
Formula Weight: 229.08
Melting Point: 155-156°
Beilstein Reference: 2084039
UN No.: 3261
MDL No.: MFCD00020378
Formula:
C₉H₉BrO₂ Melting Pt: 155…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020378 CAS Number: 113023-73-7 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...