cis-1,4-Diacetoxy-2-butene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-2-Butene-1,4-diol Diacetate, (Z)-But-2-ene-1,4-diyl diacetate

L13024-09 L13024-18
CAAAAL13024-09EA 76 CAD
CAAAAL13024-09 CAAAAL13024-18
cis-1,4-Diacetoxy-2-butene 96%
cis-1,4-Diacetoxy-2-butene

Purity: 96% CAS No.: 25260-60-0 Molecular Formula: C8H12O4 Formula Weight: 172.18 Flash Point: 110°(230°F) Refractive Index: 1.4416 Boiling Point: 120-121°/18mm Beilstein Reference: 1724379 MDL No.: MFCD00059339

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 120…121 °C/18 mm
Density: 1.090
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059339
CAS Number: 25260-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR