Methyl hydrogen isophthalate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Isophthalic acid monomethyl ester, 3-(Methoxycarbonyl)benzoic acid, Benzene-1,3-dicarboxylic acid monomethyl ester, mono-Methyl isophthalate
L12992-03
L12992-06
CAAAAL12992-03EA
32.55
CAD
CAAAAL12992-03
CAAAAL12992-06
Methyl hydrogen isophthalate 97%
Methyl hydrogen isophthalate
Purity: 97%
CAS No.: 1877-71-0
Molecular Formula: C9H8O4
Formula Weight: 180.16
Melting Point: 193-194°
Beilstein Reference: 2048750
MDL No.: MFCD00029972
Formula:
C₉H₈O₄ MW: 180.16 g/mol Melting Pt: 193…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00029972 CAS Number: 1877-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...