3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
Supplier: Thermo Scientific Chemicals
L12958-14
L12958-22
CAAAAL12958-14EA
43.45
CAD
CAAAAL12958-14
CAAAAL12958-22
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane
Purity: 97%
CAS No.: 1455-42-1
Molecular Formula: C15H28O6
Formula Weight: 304.39
Melting Point: 197-200°
Beilstein Reference: 1574570
MDL No.: MFCD00059794
Formula:
C₁₅H₂₈O₆ Melting Pt: 197…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059794 CAS Number: 1455-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...