4-Phenyl-1-butyne 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12946-06
L12946-14
CAAAAL12946-06EA
162.74
CAD
CAAAAL12946-06
CAAAAL12946-14
4-Phenyl-1-butyne 98%
4-Phenyl-1-butyne
Purity: 98%
CAS No.: 16520-62-0
Molecular Formula: C10H10
Formula Weight: 130.19
Refractive Index: 1.5210
Boiling Point: 189-190°
MDL No.: MFCD00041667
Formula:
C₁₀H₁₀ MW: 130.19 g/mol Boiling Pt: 189…190 °C Density: 0.926 Storage Temperature: Ambient |
MDL Number:
MFCD00041667 CAS Number: 16520-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...